Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drh-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059760.6 | NP_492161.3 | 3 - 4 | 359 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |