Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026460.3 | NP_001021631.1 | 9 - 10 | 2862 | 108 | |
NM_001026461.4 | NP_001021632.1 | 9 - 10 | 2860 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |