Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026376.5 | NP_001021547.1 | 1 - 2 | 181 | 135 | |
NM_001347280.1 | NP_001334202.1 | 1 - 2 | 181 | 135 | ||
NM_059816.7 | NP_492217.2 | 1 - 2 | 187 | 135 | ||
GenBank mRNA | AF013950.1 | - | 1 - 2 | 181 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |