Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025825.5 | NP_001020996.1 | 4 - 5 | 535 | 78 | |
NM_001136277.4 | NP_001129749.1 | 3 - 4 | 379 | 78 | ||
NM_059825.7 | NP_492226.2 | 4 - 5 | 537 | 78 | ||
GenBank mRNA | AJ133374.1 | - | 4 - 5 | 532 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |