Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gon-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129034.3 | NP_001122506.1 | 12 - 13 | 2545 | 143 | |
NM_001136316.4 | NP_001129788.1 | 6 - 7 | 1510 | 143 | ||
NM_001373178.2 | NP_001359707.1 | 14 - 15 | 2696 | 143 | ||
NM_059914.7 | NP_492315.4 | 15 - 16 | 2769 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |