Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsr-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026439.3 | NP_001021610.1 | 3 - 4 | 156 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |