Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cel-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025808.5 | NP_001020979.1 | 10 - 11 | 1592 | 83 | |
NM_001025809.2 | NP_001020980.1 | 10 - 11 | 1534 | 83 | ||
GenBank mRNA | AF003925.1 | - | 10 - 11 | 1439 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |