Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060215.3 | NP_492616.1 | 7 - 8 | 749 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |