Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mut-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060259.6 | NP_492660.3 | 6 - 7 | 1188 | 98 | |
NM_060260.9 | NP_492661.3 | 6 - 7 | 1176 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |