Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ztf-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373713.1 | NP_001360315.1 | 2 - 3 | 363 | 120 | |
NM_001373725.1 | NP_001359947.1 | 2 - 3 | 363 | 120 | ||
NM_001381400.1 | NP_001368396.1 | 2 - 3 | 213 | 120 | ||
NM_001381421.1 | NP_001368397.1 | 2 - 3 | 213 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |