Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: haf-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026495.5 | NP_001021666.1 | 2 - 3 | 336 | 57 | |
NM_001306255.3 | NP_001293184.1 | 1 - 2 | 56 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |