Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: deps-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306264.3 | NP_001293193.1 | 10 - 11 | 1650 | 71 | |
NM_058341.6 | NP_490742.1 | 11 - 12 | 1789 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |