Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nlp-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058507.7 | NP_490908.1 | 2 - 3 | 261 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |