Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nekl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306391.3 | NP_001293320.1 | 8 - 9 | 1267 | 81 | |
NM_001306392.3 | NP_001293321.1 | 7 - 8 | 1153 | 81 | ||
NR_131355.1 | 8 - 9 | 1267 | 81 | |||
NR_131356.1 | 7 - 8 | 1156 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |