Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptr-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026388.5 | NP_001021559.2 | 11 - 12 | 2358 | 72 | |
NR_131361.1 | 11 - 12 | 2911 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |