Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129055.4 | NP_001122527.1 | 7 - 8 | 932 | 115 | |
NM_001313513.3 | NP_001300442.1 | 6 - 7 | 417 | 115 | ||
NM_001313514.3 | NP_001300443.1 | 6 - 7 | 873 | 115 | ||
NM_058736.5 | NP_491137.2 | 6 - 7 | 414 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |