Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025843.3 | NP_001021014.1 | 4 - 5 | 800 | 78 | |
NM_001025845.4 | NP_001021016.1 | 4 - 5 | 800 | 78 | ||
NM_001375250.2 | NP_001361962.1 | 4 - 5 | 802 | 78 | ||
NM_001375251.3 | NP_001361960.1 | 4 - 5 | 800 | 78 | ||
NM_001375252.3 | NP_001361961.1 | 4 - 5 | 802 | 78 | ||
NM_058826.2 | NP_491227.1 | 4 - 5 | 800 | 78 | ||
NM_058829.5 | NP_491230.1 | 4 - 5 | 785 | 78 | ||
GenBank mRNA | AF144257.1 | - | 4 - 5 | 767 | 78 | |
AF144258.1 | - | 4 - 5 | 767 | 78 | ||
AF144259.1 | - | 4 - 5 | 767 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |