Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375250.2 | NP_001361962.1 | 6 - 7 | 1417 | 88 | |
NM_058826.2 | NP_491227.1 | 6 - 7 | 1415 | 88 | ||
NM_058829.5 | NP_491230.1 | 6 - 7 | 1400 | 88 | ||
GenBank mRNA | AF144257.1 | - | 6 - 7 | 1382 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |