Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026325.2 | NP_001021496.1 | 3 - 4 | 391 | 95 | |
NM_001026326.4 | NP_001021497.1 | 3 - 4 | 279 | 95 | ||
NM_001381834.2 | NP_001367474.1 | 3 - 4 | 279 | 95 | ||
GenBank mRNA | AF048834.1 | - | 3 - 4 | 391 | 95 | |
AF048835.1 | - | 3 - 4 | 391 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |