Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nab-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379751.2 | NP_001367535.1 | 6 - 7 | 790 | 90 | |
NM_001379754.2 | NP_001367538.1 | 5 - 6 | 599 | 90 | ||
NM_001382891.1 | 4 - 5 | 981 | 90 | |||
NM_001382892.1 | 6 - 7 | 791 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |