Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndx-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058935.7 | NP_491336.2 | 4 - 5 | 693 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |