Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356817.3 | NP_001343707.1 | 17 - 18 | 4575 | 91 | |
NM_001356818.3 | NP_001343735.1 | 6 - 7 | 1667 | 91 | ||
NM_058952.5 | NP_491353.2 | 17 - 18 | 4596 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |