Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001262699.2 | NP_001249628.1 | 2 - 3 | 295 | 92 | |
NM_001372632.2 | NP_001360115.1 | 2 - 3 | 288 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |