Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lfe-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025877.6 | NP_001021048.1 | 2 - 3 | 361 | 72 | |
NM_001025878.7 | NP_001021049.1 | 5 - 6 | 473 | 72 | ||
NM_001262849.3 | NP_001249778.1 | 5 - 6 | 456 | 72 | ||
NM_001372686.1 | NP_001359911.1 | 4 - 5 | 395 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |