Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mat-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026543.7 | NP_001021714.1 | 11 - 12 | 2309 | 100 | |
NM_001377581.3 | NP_001364721.1 | 10 - 11 | 2232 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |