Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001262958.3 | NP_001249887.1 | 13 - 14 | 1480 | 107 | |
NM_001383090.1 | 13 - 14 | 1482 | 107 | |||
NM_059185.7 | NP_491586.1 | 12 - 13 | 1458 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |