Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059285.4 | NP_491686.1 | 7 - 8 | 1822 | 115 | |
GenBank mRNA | AF116529.1 | - | 7 - 8 | 1844 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |