Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025795.3 | NP_001020966.2 | 3 - 4 | 305 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |