Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: avr-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025792.4 | NP_001020963.1 | 11 - 12 | 1143 | 109 | |
NM_001383179.1 | 16 - 17 | 1688 | 109 | |||
NM_001383181.1 | 14 - 15 | 1860 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |