Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026159.6 | NP_001021330.1 | 4 - 5 | 560 | 107 | |
NM_171391.7 | NP_741456.1 | 3 - 4 | 625 | 107 | ||
NR_131580.1 | 3 - 4 | 506 | 107 | |||
GenBank mRNA | AF273776.1 | - | 3 - 4 | 615 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |