Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pash-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306532.2 | NP_001293461.1 | 9 - 10 | 1370 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |