Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028998.4 | NP_001024169.1 | 9 - 10 | 1597 | 71 | |
NM_001028999.5 | NP_001024170.1 | 9 - 10 | 1948 | 71 | ||
NM_001029002.4 | NP_001024173.1 | 8 - 9 | 1470 | 71 | ||
NM_001029004.5 | NP_001024175.1 | 8 - 9 | 1371 | 71 | ||
NM_001313278.3 | NP_001300207.1 | 8 | 1359 | 71 | ||
NM_001380634.1 | NP_001367924.1 | 9 - 10 | 1585 | 71 | ||
NM_001380635.1 | NP_001367923.1 | 9 - 10 | 1587 | 71 | ||
NM_001383298.1 | 8 - 9 | 2287 | 71 | |||
GenBank mRNA | AF427474.1 | - | 9 - 10 | 1607 | 71 | |
AF427475.1 | - | 9 - 10 | 1619 | 71 | ||
AF427476.1 | - | 9 - 10 | 1790 | 71 | ||
AF427477.1 | - | 9 - 10 | 1802 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |