Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072253.5 | NP_504654.1 | 9 - 10 | 1439 | 121 |
Gene Symbol: ubh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307792.3 | NP_001294721.2 | 3 - 4 | 555 | 121 |
Gene Symbol: ubh-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072252.7 | NP_504653.2 | 3 - 4 | 555 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |