Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028557.3 | NP_001023728.1 | 1 - 2 | 129 | 123 | |
GenBank mRNA | AY532645.1 | - | 1 - 2 | 120 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |