Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136431.4 | NP_001129903.1 | 14 - 15 | 11862 | 83 | |
NM_001269453.3 | NP_001256382.1 | 12 - 13 | 11613 | 83 | ||
NM_171544.7 | NP_741632.2 | 15 - 16 | 12286 | 83 | ||
GenBank mRNA | AF053496.1 | - | 14 - 15 | 11862 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |