Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270068.6 | NP_001256997.2 | 17 - 18 | 2438 | 141 | |
NM_001313456.4 | NP_001300385.1 | 18 | 2453 | 141 | ||
NM_001313457.4 | NP_001300386.1 | 3 - 4 | 512 | 141 | ||
NM_171645.6 | NP_741740.3 | 18 - 19 | 2569 | 141 | ||
GenBank mRNA | EU729334.1 | - | 17 - 18 | 2420 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |