Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270068.6 | NP_001256997.2 | 7 - 8 | 1016 | 75 | |
NM_001313456.4 | NP_001300385.1 | 7 | 998 | 75 | ||
NM_171645.6 | NP_741740.3 | 7 - 8 | 1015 | 75 | ||
NM_171646.5 | NP_741741.2 | 7 - 8 | 1017 | 75 | ||
GenBank mRNA | EU729334.1 | - | 7 - 8 | 998 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |