Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375019.2 | NP_001361932.1 | 12 | 4584 | 154 | |
NM_001375020.2 | NP_001362134.1 | 11 - 12 | 4582 | 154 | ||
NM_001375021.2 | NP_001361933.1 | 14 - 15 | 6132 | 154 | ||
NM_001375022.2 | NP_001362133.1 | 14 - 15 | 6134 | 154 | ||
NM_001375024.2 | NP_001362136.1 | 13 - 14 | 6059 | 154 | ||
NM_001383564.1 | 14 - 15 | 6130 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |