Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375021.2 | NP_001361933.1 | 2 - 3 | 167 | 60 | |
NM_001375022.2 | NP_001362133.1 | 2 - 3 | 163 | 60 | ||
NM_001375024.2 | NP_001362136.1 | 1 - 2 | 88 | 60 | ||
NM_001383564.1 | 2 - 3 | 163 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |