Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029293.3 | NP_001024464.1 | 11 - 12 | 1628 | 87 | |
NM_001029296.8 | NP_001024467.1 | 11 - 12 | 1512 | 87 | ||
GenBank mRNA | AF083643.1 | - | 11 - 12 | 1436 | 87 | |
AF083647.1 | - | 11 - 12 | 1628 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |