Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171742.5 | NP_741860.2 | 2 - 3 | 376 | 63 | |
NM_171743.6 | NP_741861.2 | 2 - 3 | 378 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |