Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171756.4 | NP_741878.1 | 1 - 2 | 46 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |