Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029268.5 | NP_001024439.1 | 11 - 12 | 1442 | 79 | |
NM_171796.5 | NP_741925.2 | 7 - 8 | 1285 | 79 | ||
GenBank mRNA | AJ133838.1 | - | 7 - 8 | 1010 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |