Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eor-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029299.6 | NP_001024470.1 | 4 - 5 | 550 | 92 | |
NM_001029300.6 | NP_001024471.1 | 4 - 5 | 550 | 92 | ||
GenBank mRNA | AF502570.1 | - | 4 - 5 | 497 | 92 | |
AF519109.1 | - | 4 - 5 | 475 | 92 | ||
AF519110.1 | - | 4 - 5 | 475 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |