Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eor-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029299.6 | NP_001024470.1 | 7 - 8 | 1171 | 58 | |
GenBank mRNA | AF502570.1 | - | 7 - 8 | 1118 | 58 | |
AF519109.1 | - | 7 - 8 | 1096 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |