Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171848.6 | NP_740978.1 | 8 - 9 | 1261 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |