Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gar-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029230.3 | NP_001024401.1 | 14 - 15 | 1682 | 78 | |
NM_001029231.3 | NP_001024402.1 | 15 - 16 | 1845 | 78 | ||
GenBank mRNA | AF075245.1 | - | 14 - 15 | 1682 | 78 | |
AF117300.1 | - | 15 - 16 | 1775 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |