Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ajm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330908.3 | NP_001317889.1 | 17 - 18 | 4081 | 61 | |
NM_001330909.3 | NP_001317890.1 | 17 - 18 | 3856 | 61 | ||
NM_001330910.3 | NP_001317891.1 | 15 - 16 | 3724 | 61 | ||
NM_001330911.3 | NP_001317888.1 | 14 - 15 | 3610 | 61 | ||
NM_001330912.3 | NP_001317897.1 | 13 - 14 | 3294 | 61 | ||
NM_001330913.3 | NP_001317898.1 | 12 - 13 | 3049 | 61 | ||
NM_001330914.3 | NP_001317899.1 | 11 - 12 | 2977 | 61 | ||
NM_001330915.3 | NP_001317736.1 | 9 - 10 | 2680 | 61 | ||
NM_001381045.1 | NP_001368606.1 | 23 - 24 | 4864 | 61 | ||
NM_001381046.2 | NP_001367383.1 | 22 - 23 | 4749 | 61 | ||
NM_001381048.1 | NP_001367381.1 | 23 - 24 | 4901 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |