Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tdo-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065883.5 | NP_498284.1 | 3 - 4 | 514 | 83 | |
NR_136366.1 | 3 - 4 | 510 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |