Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pde-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047333.6 | NP_001040798.1 | 9 - 10 | 1384 | 107 | |
NM_001306543.3 | NP_001293472.1 | 9 - 10 | 1237 | 107 | ||
NM_001383892.1 | 9 - 10 | 1297 | 107 | |||
NM_063199.6 | NP_495600.1 | 9 - 10 | 1319 | 107 | ||
NM_063200.4 | NP_495601.1 | 7 - 8 | 1417 | 107 | ||
NM_182144.6 | NP_871944.1 | 10 - 11 | 1997 | 107 | ||
NM_182145.5 | NP_871945.1 | 7 - 8 | 1490 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |